missing translation for 'onlineSavingsMsg'
Learn More
Ethyl (E)-2-cyano-3-ethoxycrotonate, 98%, Thermo Scientific Chemicals Product Code.: 11362259

Ethyl (E)-2-cyano-3-ethoxycrotonate, 98%, Thermo Scientific Chemicals

Product Code. 11362259
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11362259

Brand: Thermo Scientific Alfa Aesar H27244.14

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932750-29-3
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD01743485
InChI Key YKXFOGAYPIPTKF-BQYQJAHWSA-N
Synonym ethyl 2e-2-cyano-3-ethoxybut-2-enoate, ethyl e-2-cyano-3-ethoxycrotonate, ethyl 2-cyano-3-ethoxybut-2-enoate, 2-butenoic acid, 2-cyano-3-ethoxy-, ethyl ester, ethyl e-2-cyano-3-ethoxybut-2-enoate, e-ethyl 2-cyano-3-ethoxybut-2-enoate, ethyl e-2-cyano-3-ethoxy-2-butenoate, e-2-cyano-3-ethoxycrotonic acid ethyl ester, e-2-cyano-3-ethoxy-2-butenoic acid ethyl esterShow MoreShow Less
PubChem CID 6387109
IUPAC Name ethyl (E)-2-cyano-3-ethoxybut-2-enoate
SMILES CCOC(=C(C#N)C(=O)OCC)C

Specifications

Melting Point 71°C to 73°C
Quantity 25 g
UN Number UN3439
Formula Weight 183.21
Percent Purity 98%
Chemical Name or Material Ethyl (E)-2-cyano-3-ethoxycrotonate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243142 Certificate TypeCertificate of Analysis Date02/09/2022 Catalog NumberH27244.14,H27244.06
Lot Number10237800 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH27244.14,H27244.06
Lot Number10155806 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27244.14,H27244.06
Lot Number10147859 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27244.14,H27244.06
Lot Number10228963 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27244.14,H27244.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl (E)-2-cyano-3-ethoxycrotonate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.