Learn More
FCGR2A Mouse anti-Human, Clone: OTI15G5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF500654

Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.Especificaciones
FCGR2A | |
Monoclonal | |
Unconjugated | |
FCGR2A | |
CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A1, FcGR, IGFR2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2212 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
OTI15G5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P12318 | |
FCGR2A | |
Full length human recombit protein of human FCGR2A produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Sugerencias de productos
Customers who viewed this item also viewed.
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.