missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Mesothelin APC-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
Marca: R&D Systems FAB32652A
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Mesothelin Monoclonal specifically detects Mesothelin in Human samples. It is validated for Flow Cytometry.
Especificaciones
Mesothelin | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10232 | |
Human | |
Purified |
Flow Cytometry | |
420411 | |
Flow Cytometry 10 uL/10^6 cells | |
AAH09272 | |
MSLN | |
Mouse myeloma cell line NS0-derived recombinant human Mesothelin Glu296-Gly580 Accession # AAH09272 | |
100 Tests | |
Primary | |
Detects human Mesothelin in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Human Mesothelin APC-conjugated Antibody, R&D Systems™ > 100 Tests; APC
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido