Learn More
Phospho-Caveolin 2 (Tyr19) Polyclonal Antibody, Alexa Fluor™ 488, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA1060A488
Descripción
PA1-060 detects phospho-caveolin-2 Y19 from human and mouse samples. Immunofluorescent staining of phospho-caveolin-2 Y19 in NIH 3T3 cells using PA1-060-A488 results in a distinct punctate staining pattern at the cell periphery.
Caveolae are specialized domains of the plasma membrane that are implicated in the sequestration of a variety of lipid and protein molecules. It has been suggested that these important cellular organelles have a pivotal role in such diverse biochemical processes as lipid metabolism, growth regulation, signal transduction, and apoptosis. Caveolin interacts with and regulates heterotrimeric G-proteins. Currently, there are three members of the caveolin multigene family which are known to encode 21-24 kDa integral membrane proteins that comprise the major structural component of the caveolar membrane in vivo. Caveolin-2 protein is abundantly expressed in fibroblasts and differentiated adipocytes, smooth and skeletal muscle, and endothelial cells. The expression of caveolin-1 is similar to that of caveolin-2 while caveolin-3 expression appears to be limited to muscle tissue types.
Especificaciones
Phospho-Caveolin 2 (Tyr19) | |
Polyclonal | |
Alexa Fluor 488 | |
CAV2 | |
AI447843; CAV; CAV2; caveolae; caveolae protein, 20-kD; caveolin 2; caveolin 2 isoform a and b; caveolin-2; MGC12294; Phospho-CAV2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12390, 858 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry | |
1.0 mg/mL | |
PBS with 1.0mg/mL BSA and 0.05% sodium azide | |
P51636, Q9WVC3 | |
CAV2 | |
Synthetic phosphopeptide corresponding to residues M(14) A D D A (pY) S H H S G C(25) of mouse CAV2. | |
50 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Sugerencias de productos
Customers who viewed this item also viewed.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.