Learn More
RAR gamma-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA3813
360.01 EUR hasta el 2025-03-21
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
PA3-813 detects recombinant human retinoic acid receptor (RAR) gamma-1. The immunizing sequence is 100% conserved in mouse. This antibody detects RAR gamma-1, but not RAR gamma-2, or either of the alpha or beta forms of RAR. PA3-813 has been successfully used in Western blot, immunoprecipitation and gel shift procedures. By Western blot, this antibody detects an ∽50 kDa protein representing RAR gamma-1 from transiently transfected 293T cells. PA3-813 has been shown to efficiently super shift the human RAR gamma-1/retinoid X receptor (RXR)/RAR response element complex in gel super shift experiments using a number of different response elements. PA3-813 immunizing peptide corresponds to the N-terminus of RAR gamma-1. This peptide (Cat. # PEP-228) is available for use in neutralization and control experiments.
Retinoic acid, a metabolite of vitamin A, is necessary for normal organogenesis but acts as a teratogen at high levels during embryonic and fetal development. Retinoic acid functions through its interaction with the nuclear protein, retinoic acid receptor (RAR). RAR belongs to the steroid and thyroid hormone superfamily of nuclear receptor proteins which exert their effects by binding to specific DNA response elements, thus regulating gene expression in target cells. RAR exists as three major subtypes: alpha, beta and gamma. Two main isoforms of RAR gamma exist: RAR gamma-1 has been found to be predominantly expressed in skin, while RAR gamma-2 has been found early in embryogenesis and in embryonal carcinoma cells. RAR gamma is also known as RARD. The corresponding gene for the retinoic acid receptor gamma 1 is NR1B3.
Especificaciones
RAR gamma-1 | |
Polyclonal | |
Unconjugated | |
Rarg | |
Nr1b3; Nuclear receptor subfamily 1 group B member 3; RAR gamma 2; RAR Gamma-1; RARC; RARD; Rarg; RAR-gamma; RARgamma2; retinoic acid; retinoic acid nuclear receptor gamma variant 1; retinoic acid nuclear receptor gamma variant 2; retinoic acid receptor; retinoic acid receptor gamma; retinoic acid receptor gamma 4; retinoic acid receptor, gamma | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Gel Shift, Immunoprecipitation, Western Blot | |
Conc. Not Determined | |
whole serum diluted in PBS with 0.05% sodium azide | |
P13631, P18911 | |
Rarg | |
Synthetic peptide corresponding to the residues E(40) M L S P S F R G L G Q P D L P K E(57) of the N-terminal end of RAR gamma-1. | |
RUO | |
19411, 5916, 685072 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.