Learn More
RASA1 Monoclonal Antibody (B4F8), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA4001
Descripción
MA4-001 detects GTPase activating protein (GAP) from human, mouse, rat, bovine and non-human primate tissues. MA4-001 has been successfully used in Western blot, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody recognizes a single 120 kDa protein representing ras GAP from RS-2 cell lysate. Immunofluorescence staining of GAP in mouse fibroblast cells with MA4-001 results in diffuse cytoplasmic staining. Following the addition of PDGF, immunofluorescence staining shows that some GAP rapidly translocates to the plasma membrane. MA4-001 immunoprecipitates GAP that is complexed with the GAP related p190. The MA4-001 antigen is full length recombinant human GAP. Epitope mapping studies suggest that this antibody binds a portion of GAP that contains the src homology regions SH2 & SH3. Reconstitute with PBS.
TACC1 is located on 8p11 chromosomal region that is amplified in approximately 15% of all breast tumor samples. The short arm of chromosome 8 also contains FGFR1 whose expression is enhanced in most breast cancer tumors. TACC family members, TACC1, TACC2, and TACC3, map very closely to the corresponding FGFR1, FGFR2, FGFR3 genes on chromosomes 4,8, and 10. Subsequently, since they are phylogenetically related, it is proposed that TACC and FGFR have similar roles in cell growth and differentiation. Also, TACC1 contains a conserved C-terminal region as in the Drosophila homolog, D-TACC. It has been shown that D-TACC is necessary for normal spindle function, and the mammalian TACC proteins appears to interact with centrosomes and microtubules in a similar manner.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Especificaciones
RASA1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P09851, P20936, P50904 | |
RASA1 | |
Full length recombinant human GAP. | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
B4F8 | |
Unconjugated | |
RASA1 | |
CMAVM; CM-AVM; Gap; GAPX; GTPase-activating protein; p120; p120 RAS GTPase activating protein; p120GAP; p120RASGAP; p120-rasGAP; PKWS; Ras GTPase-activating protein 1; ras GTPase-activating protein 1-like protein; Ras p21 protein activator; RAS p21 protein activator (GTPase activating protein RAS p21); RAS p21 protein activator (GTPase activating protein) 1; RAS p21 protein activator 1; Rasa; RASA1; RASGAP; triphosphatase-activating protein; unnamed protein product | |
Mouse | |
Protein A | |
RUO | |
218397, 25676, 282032, 5921 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Sugerencias de productos
Customers who viewed this item also viewed.
Certificados
Se requiere un número de lote para mostrar resultados para certificados. Para encontrar su número de lote en pedidos anteriores, utilice el área de estado de pedidos.
Número de lote | Tipo de certificado | Fecha | Catalog Number |
---|---|---|---|
AB407936 | Certificado de análisis | 26/02/2025 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.