Learn More
Novus Biologicals™ Recombinant Human Glucose 6 phosphate isomerase His Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
Marca: Novus Biologicals™ NBP2-52232-0.1MG
Descripción
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-558 of Human Glucose 6 phosphate isomerase The Recombinant Human Glucose 6 phosphate isomerase Protein is derived from E. coli. The Recombinant Human Glucose 6 phosphate isomerase Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Especificaciones
1mg/mL | |
20mM Tris-HCl buffer (pH8.0) containing 1mM DTT, 10% glycerol | |
TMW: 65.3kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
Glucose 6 phosphate isomerase | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
SDS-PAGE | |
2821 | |
0.1 mg | |
AMFGNPI, Autocrine motility factor, DKFZp686C13233, EC 5.3.1.9, glucose phosphate isomerase, glucose-6-phosphate isomerase, hexose monophosphate isomerase, hexosephosphate isomerase, neuroleukin, NLKSA36, oxoisomerase, PGI, PHI, Phosphoglucose isomerase, phosphohexomutase, Phosphohexose isomerase, phosphosaccharomutase, SA-36, Sperm antigen 36, sperm antigen-36 | |
Specific activity is > 400 units/mg obtained by measuring the increase of NADPH absorbance at 340 nm resulting from the reduction of NADP. One unit will convert 1.0 umole of D-Fructose 6-phosphate to D-glucose 6-phosphate per minute at pH 7.4 at 37C. | |
Recombinant |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For research use only and is not approved for use in humans or in clinical diagnosis