missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human MR1 GST (N-Term) Protein
Recombinant Protein
Marca: Novus Biologicals™ H00003140-Q01-25ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Especificaciones
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
3140 | |
25 μg | |
Class I histocompatibility antigen-like protein, FLJ31593, HLALS, major histocompatibility complex class I-related gene protein, major histocompatibility complex, class I-like sequence, major histocompatibility complex, class I-related, MHC class I-like antigen MR-1, MHC class I-related gene protein, MHC class-I related-gene protein | |
Immunology | |
MR1 |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
MolecularWeight-theroretical: 36.74 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
MR1 | |
>80% by SDS-PAGE and Coomassie blue staining |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Novus Biologicals™ Recombinant Human MR1 GST (N-Term) Protein > 25 μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido