missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUOX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA558294
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: RLHVVGAPGG QSLSLSLDDL HNFPRYEITV TLQCAGNRRS EMTQVKEVKG LEWRTGAIST ARWAGARLCD VLAQAGHQLC ETEAHVCFEG LDSDPTGT Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.
Especificaciones
SUOX | |
Polyclonal | |
Unconjugated | |
SUOX | |
SO; sulfite oxidase; sulfite oxidase, mitochondrial; Suox | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6821 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P51687 | |
SUOX | |
Recombinant protein corresponding to Human SUOX | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
SUOX Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido