Learn More
Synapsin 1 Monoclonal Antibody (7H10G6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA531919
Description
MA5-31919 has been tested in indirect ELISA.
SYN1 (synapsin 1) is a member of the synapsin gene family. Synapsins encode neuronal phosphoproteins that associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains and are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. Synapsin 1 plays a role in regulation of axonogenesis and synaptogenesis. It serves as a substrate for several different protein kinases and phosphorylation may function in the regulation of this protein in the nerve terminal. Mutations in this gene may be associated with X-linked disorders with primary neuronal degeneration such as Rett syndrome. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
Synapsin 1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
O88935, P09951, P17600 | |
SYN1 | |
Purified recombinant fragment of human SYN1 (AA: 362-511) expressed in E. coli. | |
100μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
7H10G6 | |
Unconjugated | |
SYN1 | |
Brain protein 4.1; SYN1; Syn-1; SYN1a; SYN1b; Syn1-S; synapsin 1; synapsin I; synapsin Ia; synapsin Ib; synapsin-1; SYNI; truncated synapsin 1-S | |
Mouse | |
Protein G | |
RUO | |
20964, 24949, 6853 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Suggestions
Los clientes que vieron este artículo también vieron
Your input is important to us. Please complete this form to provide feedback related to the content on this product.