missing translation for 'onlineSavingsMsg'
Learn More
1-(4-Chlorophenyl)-1H-pyrrole-2-carbaldehyde, Tech., Thermo Scientific™ Product Code.: 10621313

1-(4-Chlorophenyl)-1H-pyrrole-2-carbaldehyde, Tech., Thermo Scientific™

Product Code. 10621313
10 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10621313

Brand: Thermo Scientific Maybridge SEW04034EA

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Chemical Identifiers

CAS 37560-50-2
Molecular Formula C11H8ClNO
Molecular Weight (g/mol) 205.641
MDL Number MFCD00174283
InChI Key NNQOVJLQFYROBU-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-1h-pyrrole-2-carbaldehyde, 1-4-chlorophenyl pyrrole-2-carbaldehyde, 1-p-chlorphenyl-2-pyrrolcarbaldehyd, 1-4-chloro-phenyl-1h-pyrrole-2-carbaldehy de, 1h-pyrrole-2-carboxaldehyde,1-4-chlorophenylShow MoreShow Less
PubChem CID 737212
IUPAC Name 1-(4-chlorophenyl)pyrrole-2-carbaldehyde
SMILES C1=CN(C(=C1)C=O)C2=CC=C(C=C2)Cl

Specifications

Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 205.64
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1-(4-Chlorophenyl)-1H-pyrrole-2-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(4-Chlorophenyl)-1H-pyrrole-2-carbaldehyde, Tech., Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.