missing translation for 'onlineSavingsMsg'
Learn More
1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid, Thermo Scientific™ Product Code.: 10646430

1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid, Thermo Scientific™

Product Code. 10646430
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10646430

Brand: Thermo Scientific Maybridge CC73101DA

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Chemical Identifiers

CAS 950603-35-7
Molecular Formula C9H10F3N3O2S
Molecular Weight (g/mol) 281.25
MDL Number MFCD11101440
InChI Key PWQMNLMMMCREOV-UHFFFAOYSA-N
Synonym 1-5-trifluoromethyl-1,3,4-thiadiazol-2-yl piperidine-4-carboxylic acid
PubChem CID 33589561
IUPAC Name 1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid
SMILES OC(=O)C1CCN(CC1)C1=NN=C(S1)C(F)(F)F

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 281.25
Percent Purity 97%
Chemical Name or Material 1-[5-(trifluoromethyl)-1, 3, 4-thiadiazol-2-yl]piperidine-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.