Learn More
CD55 Mouse anti-Human, PE, Clone: IA10, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 341030
Descripción
The CD55 antibody, clone IA10, is derived from the fusion of X63Ag8.653 myeloma cells with spleen cells from BALB/c mice immunized with purified DAF.
The CD55 antibody reacts with decay-accelerating factor (DAF), a GPI-anchored single chain glycoprotein of approximately 70 kilodaltons (kDa). It has been suggested that the role of DAF is to protect cells from damage by autologous complement preventing the amplification steps of the complement cascade by interfering with the assembly of the C3 and the C5 convertases. The complement components in the formation of C3 convertases include: C4b2a (classical pathway) and C3bBb (alternative pathway). In the formation of C5 convertases, the complement components include: C4b2a3b (classical pathway) and C3bBb3b (alternative pathway).

Especificaciones
CD55 | |
Monoclonal | |
25μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
50 Tests | |
Immunology | |
Human | |
IgG2a κ |
Flow Cytometry | |
IA10 | |
PE | |
Mouse | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.