Learn More
CSH1 Monoclonal Antibody (OTI1C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marca: Origene Technologies TA506814

Descripción
The protein encoded by this gene is a member of the somatotropin/prolactin family of hormones and plays an important role in growth control. The gene is located at the growth hormone locus on chromosome 17 along with four other related genes in the same transcriptional orientation; an arrangement which is thought to have evolved by a series of gene duplications. Although the five genes share a remarkably high degree of sequence identity, they are expressed selectively in different tissues. Alternative splicing generates additional isoforms of each of the five growth hormones, leading to further diversity and potential for specialization. This particular family member is expressed mainly in the placenta and utilizes multiple transcription initiation sites. Expression of the identical mature proteins for chorionic somatomammotropin hormones 1 and 2 is upregulated during development, although the ratio of 1 to 2 increases by term. Mutations in this gene result in placental lactogen deficiency and Silver-Russell syndrome.Especificaciones
CSH1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P0DML2 | |
CSH1 | |
Full length human recombit protein of human CSH1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1C9 | |
Unconjugated | |
CSH1 | |
CS-1, CSA, CSMT, GHB3, PL, hCS-1, hCS-A | |
Mouse | |
Affinity Chromatography | |
RUO | |
1442 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.