Learn More
Novus Biologicals™ Recombinant Human BAP18 His Protein
Highly purified. Generating reliable and reproducible results.
Marca:  Novus Biologicals™ NBP2-22781
Código de producto. 18117921
  • 528.00€ / 0.05 miligramo
Explore más ofertas especiales
Descripción
Descripción
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-172 of Human BAP18 The Recombinant Human BAP18 Protein is derived from E. coli. The Recombinant Human BAP18 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
NP_777553
20mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 10% glycerol, 1mM DTT
20.3kDa
SDS-PAGE
E. Coli
−20°C or −80°C. Avoid freeze-thaw cycles.
Unconjugated
Recombinant
SDS-PAGE
124944
BAP18 Protein
0.05 mg
BAP18, 1-172aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMTSASTK VGEIFSAAGA AFTKLGELTM QLHPVADSSP AGAKWTETEI EMLRAAVKRF GDDLNHISCV IKERTVAQIK ATVKRKVYED SGIPLPAESP KKGPKKVASG VLSPPPAAPP PSSSSVPEAG GPPIKKQKAD VTLSALNDSD ANSDVVDIEG LGETPPAKKL NFDQA
RUO
Human
DNA replication Transcription Translation and Splicing
Certificados
Ofertas especiales
Ofertas especiales
Título del producto
Novus Biologicals™ Recombinant Human BAP18 His Protein > 0.05mg