missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylisobutyramide, 99%, Thermo Scientific Chemicals Product Code.: 15440238

N,N-Dimethylisobutyramide, 99%, Thermo Scientific Chemicals

Product Code. 15440238
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15440238

Brand: Thermo Scientific Alfa Aesar H53430.22

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

1- chloro-N,N-2-trimethylpropenylamine readily prepared' from N,N-dimethylisobutyramide. Isoxazoles are prepared by deprotonation of N,N-dimethylisobutyramide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1- chloro-N,N-2-trimethylpropenylamine readily prepared′ from N,N-dimethylisobutyramide. Isoxazoles are prepared by deprotonation of N,N-dimethylisobutyramide.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21678-37-5
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00075378
InChI Key GXMIHVHJTLPVKL-UHFFFAOYSA-N
Synonym n,n-dimethylisobutyramide, n,n,2-trimethylpropionamide, n,n-dimethyl-isobutyramide, acmc-20ak9x, n,n-dimethylisobutylamide, isobutyric acid dimethylamide, propanamide, n,n,2-trimethyl, propanamide,n,n,2-trimethyl, n,n-dimethyl2-methylpropionamideShow MoreShow Less
PubChem CID 243415
IUPAC Name N,N,2-trimethylpropanamide
SMILES CC(C)C(=O)N(C)C

Specifications

Density 0.892
Boiling Point 175°C to 176°C (44 mmHg)
Flash Point 154°C (309°F)
Linear Formula (CH3)2CHCON(CH3)2
Refractive Index 1.439
Quantity 100 g
Beilstein 1742178
Solubility Information Soluble in water.
Formula Weight 115.18
Percent Purity 99%
Chemical Name or Material N,N-Dimethylisobutyramide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW06J059 Certificate TypeCertificate of Analysis Date13/09/2023 Catalog NumberH53430.06,H53430.14,H53430.22
Lot NumberW12E024 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53430.06,H53430.14,H53430.22
Lot NumberP28C046 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53430.06,H53430.14,H53430.22
Lot NumberR23D073 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53430.06,H53430.14,H53430.22
Lot NumberN08E041 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53430.06,H53430.14,H53430.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylisobutyramide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.