Learn More
PSMA6 Mouse anti-Canine, Human, Mouse, Rat, Clone: UMAB102, liquid, UltraMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies UM870032

Descripción
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. A pseudogene has been identified on the Y chromosome.Especificaciones
PSMA6 | |
Monoclonal | |
0.5-1.0 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P60900, P60901, Q9QUM9 | |
PSMA6 | |
Full length human recombit protein of human PSMA6 produced in E.coli. | |
30 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
UMAB102 | |
Unconjugated | |
PSMA6 | |
IOTA, PROS27, p27K | |
Mouse | |
Affinity Chromatography | |
RUO | |
26443, 29673, 480290, 5687 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Sugerencias de productos
Customers who viewed this item also viewed.
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.