Learn More
SCP2 Monoclonal Antibody (OTI3F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marca: Origene Technologies TA507255

Descripción
This gene encodes two proteins: sterol carrier protein X (SCPx) and sterol carrier protein 2 (SCP2), as a result of transcription initiation from 2 independently regulated promoters. The transcript initiated from the proximal promoter encodes the longer SCPx protein, and the transcript initiated from the distal promoter encodes the shorter SCP2 protein, with the 2 proteins sharing a common C-terminus. Evidence suggests that the SCPx protein is a peroxisome-associated thiolase that is involved in the oxidation of branched chain fatty acids, while the SCP2 protein is thought to be an intracellular lipid transfer protein. This gene is highly expressed in organs involved in lipid metabolism, and may play a role in Zellweger syndrome, in which cells are deficient in peroxisomes and have impaired bile acid synthesis. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms. The full-length nature of all transcript variants has not been determined.Especificaciones
SCP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P22307 | |
SCP2 | |
Full length human recombit protein of human SCP2 produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI3F5 | |
Unconjugated | |
SCP2 | |
NLTP, NSL-TP, SCP-2, SCP-CHI, SCP-X, SCPX | |
Mouse | |
Affinity Chromatography | |
RUO | |
6342 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.