Learn More
TACC3 Monoclonal Antibody (OTI6C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marca: Origene Technologies TA501262S

Descripción
TACC3 Monoclonal antibody specifically detects TACC3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
TACC3 | |
Monoclonal | |
0.53 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9Y6A5 | |
Mouse | |
Affinity chromatography | |
RUO | |
10460 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI6C6 | |
Unconjugated | |
TACC3 | |
Aint; Arnt interacting protein; ARNT-interacting protein; C86661; ERIC1; ERIC-1; Tacc3; transforming acidic coiled coil 3; transforming acidic coiled-coil containing protein 3; transforming acidic coiled-coil-containing protein 3; transforming, acidic coiled-coil containing protein 3 | |
Full length human recombinant protein of human TACC3 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Suggestions
Los clientes que vieron este artículo también vieron
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.