missing translation for 'onlineSavingsMsg'
Learn More
2-Fluoro-6-methoxybenzyl alcohol, 98%, Thermo Scientific™ Product Code.: 15465637

2-Fluoro-6-methoxybenzyl alcohol, 98%, Thermo Scientific™

Product Code. 15465637
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15465637

Brand: Thermo Scientific Alfa Aesar H32878.03

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253668-46-1
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.156
MDL Number MFCD00671766
InChI Key ATWUFIBZMGBKHR-UHFFFAOYSA-N
Synonym 2-fluoro-6-methoxybenzyl alcohol, 2-fluoro-6-methoxyphenyl methanol, acmc-1ato5, benzenemethanol, 2-fluoro-6-methoxy, 2-fluoranyl-6-methoxy-phenyl methanol, 2-fluoro-6-methoxyphenyl methan-1-ol, benzenemethanol, 2-fluoro-6-methoxy-9ciShow MoreShow Less
PubChem CID 2779320
IUPAC Name (2-fluoro-6-methoxyphenyl)methanol
SMILES COC1=C(C(=CC=C1)F)CO

Specifications

Melting Point 35°C to 39°C
Quantity 1 g
Formula Weight 156.15
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-methoxybenzyl alcohol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10156110 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10218786 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluoro-6-methoxybenzyl alcohol, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.