Learn More
Cytokeratin 10 Antibody (rKRT10/6923), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-20243-100ug
Descripción
Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Especificaciones
Cytokeratin 10 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
Recombinant fragment (around aa384-584) of human Cytokeratin 10 protein (exact sequence is proprietary) | |
100 μg | |
Cell Biology, Cellular Markers | |
3858 | |
Store at 4&drg;C. Do not freeze. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rKRT10/6923 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.