Learn More
PLA2G5 Monoclonal Antibody (3G1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA1830
Descripción
MA1-830 contains 100 μg of in vitro produced, protein A purified antibody (1 mg/mL) in PBS containing 1 mg/mL BSA and 0.05% sodium azide. MA1-830 detects Group V Secretory Phospholipase A2 from human and mouse samples. MA1-830 has been successfully used in Western blot procedures. By Western blot, MA1-830 detects a ∽15 kDa band representing Group V Secretory Phospholipase A2 from mouse macrophages. The MA1-830 immunogen is full length W79A human group V phospholipase A2.
This gene is a member of the secretory phospholipase A2 family. It is located in a tightly-linked cluster of secretory phospholipase A2 genes on chromosome 1. The encoded enzyme catalyzes the hydrolysis of membrane phospholipids to generate lysophospholipids and free fatty acids including arachidonic acid. It preferentially hydrolyzes linoleoyl-containing phosphatidylcholine substrates. Secretion of this enzyme is thought to induce inflammatory responses in neighboring cells. Alternatively spliced transcript variants have been found, but their full-length nature has not been determined.
Especificaciones
PLA2G5 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P39877, P97391 | |
PLA2G5 | |
Full length W79A human group V phospholipase A2. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
3G1 | |
Unconjugated | |
PLA2G5 | |
Ca2+-dependent phospholipase A2; calcium-dependent phospholipase A2; DKFZp686C2294; FRFB; group V phospholipase A2; GV-PLA2; hVPLA(2); MGC46205; Phosphatidylcholine 2-acylhydrolase 5; phospholipase A2 group V; phospholipase A2 group V; calcium-dependent phospholipase A2; phospholipase A2, group 5; phospholipase A2, group V; PLA2; PLA2-10; Pla2g5; sPLA2 | |
Mouse | |
Protein A | |
RUO | |
18784, 5322 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.