missing translation for 'onlineSavingsMsg'
Learn More
4,6-Bis(diphenylphosphino)phenoxazine, 98+%, Thermo Scientific Chemicals Product Code.: 11302429

4,6-Bis(diphenylphosphino)phenoxazine, 98+%, Thermo Scientific Chemicals

Product Code. 11302429
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11302429

Brand: Thermo Scientific Alfa Aesar H27330.03

Request Bulk or Custom Format

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261733-18-0
Molecular Formula C36H27NOP2
Molecular Weight (g/mol) 551.57
MDL Number MFCD03788937
InChI Key HSWZLYXRAOXOLL-UHFFFAOYSA-N
Synonym 4,6-bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-10h-phenoxazine, nixantphos, 10h-phenoxazine, 4,6-bis diphenylphosphino, 4,6-bis diphenylphosphanyl-10h-phenoxazine, nixanthphos, n-xantphos, acmc-209gp1, bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-phenoxazineShow MoreShow Less
PubChem CID 3255346
IUPAC Name (6-diphenylphosphanyl-10H-phenoxazin-4-yl)-diphenylphosphane
SMILES N1C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 251°C to 254°C
Quantity 1 g
Solubility Information Miscible with water.
Formula Weight 551.55
Percent Purity ≥98%
Chemical Name or Material 4,6-Bis(diphenylphosphino)phenoxazine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10225671 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27330.03,H27330.06,H27330.MD
Lot Number10220908 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27330.03,H27330.06,H27330.MD
Lot Number10184273 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27330.03,H27330.06,H27330.MD
Lot Number10175215 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27330.03,H27330.06,H27330.MD
Lot Number10157611 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27330.03,H27330.06,H27330.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,6-Bis(diphenylphosphino)phenoxazine, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.