Learn More
BOLA Distillation Thermometers
Distillation Thermometers PTFE-coated
Marca: BOLA B290-03
Descripción
- PTFE-jacketed glass thermometer, justified to an immersion depth of 50 mm.
- Length 300 mm
- Measuring range 0/+250:1°C.
Especificaciones
500-6000 mL | |
7.5 mm | |
-200 °C | |
50 mm |
FDA | |
250°C | |
No |
Sugerencias de productos
Customers who viewed this item also viewed.